missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Owl™ ADJ3 Adjustable System Accessories
Owl ADJ3 Adjustable System is the widest adjustable system perfect for sequencing and proteins.
£13.85 - £111.00
Specifications
For Use With (Equipment) | ADJ1 Giraffe™ Adjustable Gel Electrophoresis System |
---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11520315
|
Thermo Scientific™
ADJ3-65LM |
Heat Sink (65 x 35 cm) |
£111.00
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||||
11530315
|
Thermo Scientific™
ADJ3-GK |
Gasket |
£13.85
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||||
11510315
|
Thermo Scientific™
ADJ3-45LM |
Heat Sink (45 x 35 cm) |
£86.95
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||||
Description
- The height of the upper buffer chamber (UBC) may be adjusted from 20 cm to 80 cm by sliding the UBC up or down two support poles. The notched or offset glass plate of the gel sandwich presses against a silicone gasket on the UBC for a leakproof seal.
Specifications
ADJ1 Giraffe™ Adjustable Gel Electrophoresis System |