missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ ADJ3 Adjustable System Accessories

Owl ADJ3 Adjustable System is the widest adjustable system perfect for sequencing and proteins.

Brand:  Thermo Scientific™ ADJ3-45LM

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11510315

  • £86.95 / Each
Estimated Shipment: 29-07-2024
to see stock.
Add to basket

Description

Description

  • The height of the upper buffer chamber (UBC) may be adjusted from 20 cm to 80 cm by sliding the UBC up or down two support poles. The notched or offset glass plate of the gel sandwich presses against a silicone gasket on the UBC for a leakproof seal.

Specifications

Specifications

Heat Sink (45 x 35 cm)
Heat Sink
0.48 cm
17.7 in.
13.7 in.
ADJ1 Giraffe™ Adjustable Gel Electrophoresis System
0.18 in.
17.7 x 13.7 in. (45 x 35 cm)
45 cm
35 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers