Learn More
Thermo Scientific™ Owl™ ADJ3 Adjustable System Accessories
Owl ADJ3 Adjustable System is the widest adjustable system perfect for sequencing and proteins.
Brand:  Thermo Scientific™ ADJ3-GK
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11530315
  • £13.85 / Each
Estimated Shipment: 29-07-2024
to see stock.
Add to basket
Description
Description
  • The height of the upper buffer chamber (UBC) may be adjusted from 20 cm to 80 cm by sliding the UBC up or down two support poles. The notched or offset glass plate of the gel sandwich presses against a silicone gasket on the UBC for a leakproof seal.
Specifications
Specifications
Gasket
ADJ1 Giraffe™ Adjustable Gel Electrophoresis System
Gasket