Learn More
DWK Life Sciences O-ring, made of nitrile, for safety standard ground sockets
Shop All DWK Life Sciences Products
Click to view available options
Joint Size:
NS 14/23
NS 19/26
NS 24/29
NS 29/32
NS 34/35
NS 45/40
Unit Size:
Pack of 10
Description
Nitrile O-rings for DURAN™ safety ground joints together with the screw cap secure the connection between core and sleeve.
- In combination with the screw cap, the safety ground joints (core and sleeve) can be sealed tightly.
Specifications
Specifications
| For Use With (Application) | To secure the security grindings; Sealing |
| Joint Size | NS 29/32 |
| Quantity | 10/Pk. |
| Material | Nitrile |