missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nuf2 Antibody (8B7), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | Nuf2 |
|---|---|
| Clone | 8B7 |
| Dilution | Western Blot 1:500-1:2000, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30024819
|
Novus Biologicals
NBP3-26452-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30025082
|
Novus Biologicals
NBP3-26452-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nuf2 Monoclonal antibody specifically detects Nuf2 in Human samples. It is validated for Western Blot, ELISASpecifications
| Nuf2 | |
| Western Blot 1:500-1:2000, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| cancer/testis antigen 106, CDCA1hsNuf2, cell division cycle associated 1, Cell division cycle-associated protein 1, CT106, hNuf2, kinetochore protein Nuf2, NUF2, NDC80 kinetochore complex component, homolog (S. cerevisiae), NUF2RhNuf2R | |
| A synthesized peptide derived from Human Nuf2 [UniProt Q9BZD4] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 8B7 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 83540 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title