Learn More
Nuf2 Antibody (8B7), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26452-50ul
 View more versions of this product
Product Code. 30024819
  • £260.00 / 50µL
Description
Nuf2 Monoclonal antibody specifically detects Nuf2 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Nuf2
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Western Blot, ELISA
8B7
Western Blot 1:500-1:2000, ELISA
cancer/testis antigen 106, CDCA1hsNuf2, cell division cycle associated 1, Cell division cycle-associated protein 1, CT106, hNuf2, kinetochore protein Nuf2, NUF2, NDC80 kinetochore complex component, homolog (S. cerevisiae), NUF2RhNuf2R
A synthesized peptide derived from Human Nuf2 [UniProt Q9BZD4]
50 μL
Cell Cycle and Replication
83540
Store at -20 to -70C. Avoid freeze-thaw cycles.