missing translation for 'onlineSavingsMsg'
Learn More

Nitrobenzene, 99%, Thermo Scientific™

£27.40 - £159.00

Chemical Identifiers

CAS 98-95-3
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 123.111
MDL Number MFCD00007043
InChI Key LQNUZADURLCDLV-UHFFFAOYSA-N
Synonym nitrobenzol, benzene, nitro, oil of mirbane, mirbane oil, essence of mirbane, nitro-benzene, nitrobenzeen, oil of myrbane, nitrobenzen, mononitrobenzene
PubChem CID 7416
ChEBI CHEBI:27798
IUPAC Name nitrobenzene
SMILES C1=CC=C(C=C1)[N+](=O)[O-]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11423254
SDS View Documents
Thermo Scientific Alfa Aesar
A10585.0C
10,000g
£159.00
10000g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
11403254
SDS View Documents
Thermo Scientific Alfa Aesar
A10585.36
500g
£27.40
500g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
11413254
SDS View Documents
Thermo Scientific Alfa Aesar
A10585.0E
2500g
£52.10
2500g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Nitrobenzene is used as a solvent and as a starting material in the organic synthesis for the preparation of aniline, benzidine, azobenzene, nitrosobenzene, phenylhydroxylamine and acetaminophen. It is also used as a solvent for electrophilic reagents in synthetic chemistry. Further, it is used to prevent the unpleasant odor in shoe, floor and metal polishes. It acts as a precursor to rubber chemicals, azo dyes and pharmaceuticals.

Solubility
Miscible with ethanol, diethyl ether, acetone, benzene and organic solvents. Slightly miscible with water and carbon tetrachloride.

Notes
Incompatible with strong oxidizing agents, strong reducing agents and strong bases.
Specifications

Chemical Identifiers

98-95-3
123.111
LQNUZADURLCDLV-UHFFFAOYSA-N
7416
nitrobenzene
C6H5NO2
MFCD00007043
nitrobenzol, benzene, nitro, oil of mirbane, mirbane oil, essence of mirbane, nitro-benzene, nitrobenzeen, oil of myrbane, nitrobenzen, mononitrobenzene
CHEBI:27798
C1=CC=C(C=C1)[N+](=O)[O-]

Specifications

98-95-3
210°C to 212°C
C6H5NO2
MFCD00007043
507540
nitrobenzol, benzene, nitro, oil of mirbane, mirbane oil, essence of mirbane, nitro-benzene, nitrobenzeen, oil of myrbane, nitrobenzen, mononitrobenzene
LQNUZADURLCDLV-UHFFFAOYSA-N
nitrobenzene
7416
123.11
Nitrobenzene
1.196
87°C (188°F)
1.552
UN1662
14,6588
Miscible with ethanol,diethyl ether,acetone,benzene and organic solvents. Slightly miscible with water and carbon tetrachloride.
C1=CC=C(C=C1)[N+](=O)[O-]
123.111
CHEBI:27798
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H372-H351-H361-H227
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
Combustible liquid.

P201-P202-P210-P235-P260-P264b-P270-P271-P281-P301+P310-P302+P352-P304+P340-P308+P313-P311-P312-P330-P361-P363-P370+P378q-P501c

H227-H301+H311+H331-H350-H360F-H372

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITROBENZENE

missing translation for 'einecsNumber' : 202-716-0

missing translation for 'rtecsNumber' : DA6475000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.