Learn More
Nitrobenzene, 99%
Description
Nitrobenzene is used as a solvent and as a starting material in the organic synthesis for the preparation of aniline, benzidine, azobenzene, nitrosobenzene, phenylhydroxylamine and acetaminophen. It is also used as a solvent for electrophilic reagents in synthetic chemistry. Further, it is used to prevent the unpleasant odor in shoe, floor and metal polishes. It acts as a precursor to rubber chemicals, azo dyes and pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Nitrobenzene is used as a solvent and as a starting material in the organic synthesis for the preparation of aniline, benzidine, azobenzene, nitrosobenzene, phenylhydroxylamine and acetaminophen. It is also used as a solvent for electrophilic reagents in synthetic chemistry. Further, it is used to prevent the unpleasant odor in shoe, floor and metal polishes. It acts as a precursor to rubber chemicals, azo dyes and pharmaceuticals.
Solubility
Miscible with ethanol, diethyl ether, acetone, benzene and organic solvents. Slightly miscible with water and carbon tetrachloride.
Notes
Incompatible with strong oxidizing agents, strong reducing agents and strong bases.
Specifications
Specifications
| Melting Point | 5°C to 6°C |
| Density | 1.196 |
| Boiling Point | 210°C to 212°C |
| Flash Point | 87°C (188°F) |
| Refractive Index | 1.552 |
| Quantity | 10,000 g |
| UN Number | UN1662 |
| Beilstein | 507540 |
| Merck Index | 14,6588 |
| Solubility Information | Miscible with ethanol,diethyl ether,acetone,benzene and organic solvents. Slightly miscible with water and carbon tetrachloride. |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.