missing translation for 'onlineSavingsMsg'
Learn More

Nitrobenzene, 99%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar A10585.0E

Quantity 2500g

 View more versions of this product

Product Code. 11413254

  • £52.10 / 2500g
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Nitrobenzene is used as a solvent and as a starting material in the organic synthesis for the preparation of aniline, benzidine, azobenzene, nitrosobenzene, phenylhydroxylamine and acetaminophen. It is also used as a solvent for ele
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

98-95-3
123.111
LQNUZADURLCDLV-UHFFFAOYSA-N
7416
nitrobenzene
C6H5NO2
MFCD00007043
nitrobenzol, benzene, nitro, oil of mirbane, mirbane oil, essence of mirbane, nitro-benzene, nitrobenzeen, oil of myrbane, nitrobenzen, mononitrobenzene
CHEBI:27798
C1=CC=C(C=C1)[N+](=O)[O-]

Specifications

98-95-3
210°C to 212°C
C6H5NO2
MFCD00007043
UN1662
14,6588
Miscible with ethanol,diethyl ether,acetone,benzene and organic solvents. Slightly miscible with water and carbon tetrachloride.
C1=CC=C(C=C1)[N+](=O)[O-]
123.111
CHEBI:27798
99%
1.196
87°C (188°F)
1.552
2500g
507540
nitrobenzol, benzene, nitro, oil of mirbane, mirbane oil, essence of mirbane, nitro-benzene, nitrobenzeen, oil of myrbane, nitrobenzen, mononitrobenzene
LQNUZADURLCDLV-UHFFFAOYSA-N
nitrobenzene
7416
123.11
Nitrobenzene
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.