missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nectin-4/PVRL4 Antibody (6D3), Novus Biologicals™
Human Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | Nectin-4/PVRL4 |
|---|---|
| Clone | 6D3 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30025327
|
Novus Biologicals
NBP3-26115-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30026263
|
Novus Biologicals
NBP3-26115-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nectin-4/PVRL4 Monoclonal antibody specifically detects Nectin-4/PVRL4 in Human samples. It is validated for ELISASpecifications
| Nectin-4/PVRL4 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| 50% Glycerol, 0.01M PBS, PH 7.4 | |
| 81607 | |
| IgG1 | |
| Affinity purified |
| 6D3 | |
| ELISA | |
| Unconjugated | |
| Human | |
| Human | |
| EDSS1, Ig superfamily receptor LNIR, LNIRnectin 4, nectin-4, poliovirus receptor-related 4, poliovirus receptor-related protein 4, PRR4DKFZp686K05193 | |
| Recombinant Human Nectin-4/PVRL4 protein [UniProt Q96NY8] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title