missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nectin-4/PVRL4 Antibody (6D3), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-26115-50ul
This item is not returnable.
View return policy
Description
Nectin-4/PVRL4 Monoclonal antibody specifically detects Nectin-4/PVRL4 in Human samples. It is validated for ELISA
Specifications
| Nectin-4/PVRL4 | |
| Monoclonal | |
| Unconjugated | |
| 50% Glycerol, 0.01M PBS, PH 7.4 | |
| Human | |
| Affinity purified | |
| RUO | |
| 81607 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG1 |
| ELISA | |
| 6D3 | |
| ELISA | |
| EDSS1, Ig superfamily receptor LNIR, LNIRnectin 4, nectin-4, poliovirus receptor-related 4, poliovirus receptor-related protein 4, PRR4DKFZp686K05193 | |
| Recombinant Human Nectin-4/PVRL4 protein [UniProt Q96NY8] | |
| 50 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction