missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ND5 Mouse anti-Human, Clone: 1C12G8, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | ND5 |
|---|---|
| Clone | 1C12G8 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16828655
|
Proteintech
66613-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16818655
|
Proteintech
66613-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A small number of complex I subunits are the products of mitochondrial genes (subunits 1-7), while the remainder are nuclear encoded and imported from the cytoplasm. NADH dehydrogenase subunit 5 (ND5) is most likely a component of the hydrophobic protein fragment of Complex I. Mutations in the gene encodiing for ND5 are implicated in Mitochondrial neurogastrointestinal encephalomyopathy, an autosomal recessive multisystem disorder that results in dramatically elevated levels of circulating thymidine and deoxyuridine.Specifications
| ND5 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| MT ND5, MTND5, NADH dehydrogenase subunit 5, NADH5, ND5 | |
| MT-ND5 | |
| IgG2b | |
| Protein A | |
| Antibody |
| 1C12G8 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P03915 | |
| 4540 | |
| Peptide | |
| Primary | |
| -20°C | |
| MT-ND5 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title