missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ND5 Mouse anti-Human, Clone: 1C12G8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66613-1-IG-20UL
This item is not returnable.
View return policy
Description
A small number of complex I subunits are the products of mitochondrial genes (subunits 1-7), while the remainder are nuclear encoded and imported from the cytoplasm. NADH dehydrogenase subunit 5 (ND5) is most likely a component of the hydrophobic protein fragment of Complex I. Mutations in the gene encodiing for ND5 are implicated in Mitochondrial neurogastrointestinal encephalomyopathy, an autosomal recessive multisystem disorder that results in dramatically elevated levels of circulating thymidine and deoxyuridine.Specifications
| ND5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P03915 | |
| MT-ND5 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 1C12G8 | |
| Unconjugated | |
| MT-ND5 | |
| MT ND5, MTND5, NADH dehydrogenase subunit 5, NADH5, ND5 | |
| Mouse | |
| Protein A | |
| RUO | |
| 4540 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction