missing translation for 'onlineSavingsMsg'
Learn More

ND5 Mouse anti-Human, Clone: 1C12G8, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66613-1-IG-150UL

 View more versions of this product

Product Code. 16818655

  • £349.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

A small number of complex I subunits are the products of mitochondrial genes (subunits 1-7), while the remainder are nuclear encoded and imported from the cytoplasm. NADH dehydrogenase subunit 5 (ND5) is most likely a component of the hydrophobic protein fragment of Complex I. Mutations in the gene encodiing for ND5 are implicated in Mitochondrial neurogastrointestinal encephalomyopathy, an autosomal recessive multisystem disorder that results in dramatically elevated levels of circulating thymidine and deoxyuridine.
Specifications

Specifications

ND5
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P03915
MT-ND5
Peptide
150 μL
Primary
Human
Antibody
IgG2b
Western Blot
1C12G8
Unconjugated
MT-ND5
MT ND5, MTND5, NADH dehydrogenase subunit 5, NADH5, ND5
Mouse
Protein A
RUO
4540
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.