missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NCS1 Recombinant Rabbit Monoclonal Antibody (24GB25)

Rabbit Recombinant Monoclonal Antibody

£172.00 - £488.00

Specifications

Antigen NCS1
Clone 24GB25
Concentration 0.8 mg/mL
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30274892
View Documents
Invitrogen™
MA557504
100 μL
£488.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30276546
View Documents
Invitrogen™
MA557505
20 μL
£172.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NCS1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

This gene is a member of the neuronal calcium sensor gene family, which encode calcium-binding proteins expressed predominantly in neurons. The protein encoded by this gene regulates G protein-coupled receptor phosphorylation in a calcium-dependent manner and can substitute for calmodulin. The protein is associated with secretory granules and modulates synaptic transmission and synaptic plasticity. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

NCS1
0.8 mg/mL
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P62166, P62168, Q8BNY6
14299, 23413, 65153
A synthesized peptide derived from human NCS1 (aa 150-190).
Antibody
24GB25
-20°C
Recombinant Monoclonal
Liquid
IgG
Human, Mouse, Rat
Ncs1
9430075O15Rik; A730032G13Rik; AI836659; FLUP; Freq; frequenin; Frequenin homolog; frequenin-like protein; Frequenin-like ubiquitous protein; Mfreq; Ncs1; NCS-1; Neuronal calcium sensor 1; neuronal calcium sensor-1
Ncs1
Primary
Affinity chromatography
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.