Learn More
Invitrogen™ NCS1 Recombinant Rabbit Monoclonal Antibody (24GB25)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557504
 View more versions of this product
Product Code. 30274892
  • £488.00 / 100µL
Description
NCS1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene is a member of the neuronal calcium sensor gene family, which encode calcium-binding proteins expressed predominantly in neurons. The protein encoded by this gene regulates G protein-coupled receptor phosphorylation in a calcium-dependent manner and can substitute for calmodulin. The protein is associated with secretory granules and modulates synaptic transmission and synaptic plasticity. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NCS1
Recombinant Monoclonal
0.8 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P62166, P62168, Q8BNY6
Ncs1
A synthesized peptide derived from human NCS1 (aa 150-190).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB25
Unconjugated
Ncs1
9430075O15Rik; A730032G13Rik; AI836659; FLUP; Freq; frequenin; Frequenin homolog; frequenin-like protein; Frequenin-like ubiquitous protein; Mfreq; Ncs1; NCS-1; Neuronal calcium sensor 1; neuronal calcium sensor-1
Rabbit
Affinity chromatography
RUO
14299, 23413, 65153
-20°C