missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NCS1 Recombinant Rabbit Monoclonal Antibody (24GB25)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557505
This item is not returnable.
View return policy
Description
NCS1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene is a member of the neuronal calcium sensor gene family, which encode calcium-binding proteins expressed predominantly in neurons. The protein encoded by this gene regulates G protein-coupled receptor phosphorylation in a calcium-dependent manner and can substitute for calmodulin. The protein is associated with secretory granules and modulates synaptic transmission and synaptic plasticity. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| NCS1 | |
| Recombinant Monoclonal | |
| 0.8 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P62166, P62168, Q8BNY6 | |
| Ncs1 | |
| A synthesized peptide derived from human NCS1 (aa 150-190). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB25 | |
| Unconjugated | |
| Ncs1 | |
| 9430075O15Rik; A730032G13Rik; AI836659; FLUP; Freq; frequenin; Frequenin homolog; frequenin-like protein; Frequenin-like ubiquitous protein; Mfreq; Ncs1; NCS-1; Neuronal calcium sensor 1; neuronal calcium sensor-1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14299, 23413, 65153 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction