missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nbs1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£274.00 - £464.00
Specifications
| Dilution | Western Blot 1:1000. Use reported in multiple pieces of scientific literature, Chromatin Immunoprecipitation reported in multiple pieces of scientific literature, Flow Cytometry reported in multiple pieces of scientific literature, ELISA, Immunohistochemistry 1:100 - 1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200. Use reported in multiple pieces of scientific literature, Immunoprecipitation 3 ul. Use reported in multiple pieces of scientific literature, Immunohistochemistry-Paraffin 1:100 - 1:200, Immunoblotting, In-situ Hybridization reported in scientific literature (PMID 26415217), SDS-Page, Chromatin Immunoprecipitation (ChIP), Knockout Validated, KnockDown Validated |
|---|---|
| Target Species | Mouse, Hamster, Mammalia |
| Formulation | Whole antisera with 0.02% Sodium Azide |
| Gene Alias | ATV, AT-V1, AT-V2, Cell cycle regulatory protein p95, FLJ10155, MGC87362, NBS, NBS1P95, nibrin, Nijmegen breakage syndrome 1 (nibrin), Nijmegen breakage syndrome protein 1, p95, p95 protein of the MRE11/RAD50 complex |
| Gene Symbols | NBN |
| Product Code | Brand | dimQuantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | dimQuantity | Price | Quantity & Availability | |||||
|
18790144
|
Novus Biologicals
NB100-143SS |
£274.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18748323
|
Novus Biologicals
NB100-143 |
£464.00
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Nbs1 Polyclonal specifically detects Nbs1 in Human, Mouse, Hamster, Mammal, Primate samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, In-situ Hybridization, SDS-Page, Knockout Validated.Specifications
| Western Blot 1:1000. Use reported in multiple pieces of scientific literature, Chromatin Immunoprecipitation reported in multiple pieces of scientific literature, Flow Cytometry reported in multiple pieces of scientific literature, ELISA, Immunohistochemistry 1:100 - 1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200. Use reported in multiple pieces of scientific literature, Immunoprecipitation 3 ul. Use reported in multiple pieces of scientific literature, Immunohistochemistry-Paraffin 1:100 - 1:200, Immunoblotting, In-situ Hybridization reported in scientific literature (PMID 26415217), SDS-Page, Chromatin Immunoprecipitation (ChIP), Knockout Validated, KnockDown Validated | |
| Whole antisera with 0.02% Sodium Azide | |
| NBN |
| Mouse, Hamster, Mammalia | |
| ATV, AT-V1, AT-V2, Cell cycle regulatory protein p95, FLJ10155, MGC87362, NBS, NBS1P95, nibrin, Nijmegen breakage syndrome 1 (nibrin), Nijmegen breakage syndrome protein 1, p95, p95 protein of the MRE11/RAD50 complex | |
| Unpurified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title