Learn More
Nbs1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-143
This item is not returnable.
View return policy
Description
Nbs1 Polyclonal specifically detects Nbs1 in Human, Mouse, Hamster, Mammal, Primate samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, In-situ Hybridization, SDS-Page, Knockout Validated.
Specifications
| Rabbit | |
| RUO |