Learn More
Nbs1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-143SS
This item is not returnable.
View return policy
Description
Nbs1 Polyclonal specifically detects Nbs1 in Human, Mouse, Hamster, Mammal, Primate samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, In-situ Hybridization, SDS-Page, Knockout Validated.
Specifications
| Western Blot 1:1000. Use reported in multiple pieces of scientific literature, Chromatin Immunoprecipitation reported in multiple pieces of scientific literature, Flow Cytometry reported in multiple pieces of scientific literature, ELISA, Immunohistochemistry 1:100 - 1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200. Use reported in multiple pieces of scientific literature, Immunoprecipitation 3 ul. Use reported in multiple pieces of scientific literature, Immunohistochemistry-Paraffin 1:100 - 1:200, Immunoblotting, In-situ Hybridization reported in scientific literature (PMID 26415217), SDS-Page, Chromatin Immunoprecipitation (ChIP), Knockout Validated, KnockDown Validated | |
| ATV, AT-V1, AT-V2, Cell cycle regulatory protein p95, FLJ10155, MGC87362, NBS, NBS1P95, nibrin, Nijmegen breakage syndrome 1 (nibrin), Nijmegen breakage syndrome protein 1, p95, p95 protein of the MRE11/RAD50 complex | |
| Unpurified |
| Whole antisera with 0.02% Sodium Azide | |
| NBN | |
| Mouse, Hamster, Mammalia |