missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NAP1L1 Recombinant Rabbit Monoclonal Antibody (24GB550)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£172.00 - £488.00

Specifications

Antigen NAP1L1
Clone 24GB550
Concentration 0.415 mg/mL
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30275813
GREENER_CHOICE
View Documents
Invitrogen™
MA557494
100 μL
£488.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30275118
View Documents
Invitrogen™
MA557495
20 μL
£172.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NAP1L1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

NAP1L1 is a member of the nucleosome assembly protein (NAP) family. This protein participates in DNA replication and may play a role in modulating chromatin formation and contribute to the regulation of cell proliferation.
Specifications

Specifications

NAP1L1
0.415 mg/mL
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P28656, P55209, Q9Z2G8
4673, 53605, 89825
A synthesized peptide derived from human NAP1L1 (aa 1-55).
Antibody
24GB550
-20°C
Recombinant Monoclonal
Liquid
IgG
Human, Mouse, Rat
Nap1l1
AA407126; AI256722; Brain protein DN38; D10Ertd68e; hNRP; HSP22-like protein interacting protein; NAP1; NAP-1; NAP1L; NAP1L1; NAP-1-related protein; NRP; nucleosome assembly protein 1 like 1; nucleosome assembly protein 1-like 1; nucleosome assembly protein-1
Nap1l1
Primary
Affinity chromatography
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.