missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NAP1L1 Recombinant Rabbit Monoclonal Antibody (24GB550)

Description
NAP1L1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
NAP1L1 is a member of the nucleosome assembly protein (NAP) family. This protein participates in DNA replication and may play a role in modulating chromatin formation and contribute to the regulation of cell proliferation.
Specifications
Specifications
| Antigen | NAP1L1 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 24GB550 |
| Concentration | 0.415 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Nap1l1 |
| Gene Accession No. | P28656, P55209, Q9Z2G8 |
| Gene Alias | AA407126; AI256722; Brain protein DN38; D10Ertd68e; hNRP; HSP22-like protein interacting protein; NAP1; NAP-1; NAP1L; NAP1L1; NAP-1-related protein; NRP; nucleosome assembly protein 1 like 1; nucleosome assembly protein 1-like 1; nucleosome assembly protein-1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?