missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NAP1L1 Recombinant Rabbit Monoclonal Antibody (24GB550)

Product Code. 30275118
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30275118 20 μL 20µL
30275813 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30275118 Supplier Invitrogen™ Supplier No. MA557495

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

NAP1L1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

NAP1L1 is a member of the nucleosome assembly protein (NAP) family. This protein participates in DNA replication and may play a role in modulating chromatin formation and contribute to the regulation of cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAP1L1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 24GB550
Concentration 0.415 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Nap1l1
Gene Accession No. P28656, P55209, Q9Z2G8
Gene Alias AA407126; AI256722; Brain protein DN38; D10Ertd68e; hNRP; HSP22-like protein interacting protein; NAP1; NAP-1; NAP1L; NAP1L1; NAP-1-related protein; NRP; nucleosome assembly protein 1 like 1; nucleosome assembly protein 1-like 1; nucleosome assembly protein-1
Gene Symbols Nap1l1
Host Species Rabbit
Immunogen A synthesized peptide derived from human NAP1L1 (aa 1-55).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4673, 53605, 89825
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.