missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL1RAPL2 Antibody, R&D Systems™

Rat Monoclonal Antibody
£157.00 - £345.00
Specifications
| Antigen | IL1RAPL2 |
|---|---|
| Clone | 320017 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18749557
![]() |
R&D Systems
MAB3068-SP |
25 μg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18101841
![]() |
R&D Systems
MAB3068 |
100 μg |
£345.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL1RAPL2 Monoclonal specifically detects IL1RAPL2 in Mouse samples. It is validated for Western Blot.Specifications
| IL1RAPL2 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q9ERS6 | |
| 26280 | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL1RAPL2, Asn18-Ala358, Accession # Q9ERS6 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse IL1RAPL2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant human (rh) IL1RAPL2 is observed and no cross-reactivity with recombinant mouse (rm) IL-1 R1, rmIL-1 R2, rmIL |
| 320017 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IL-1R10, IL-1R-9, IL1R9IL-1 receptor accessory protein-like 2, IL-1R9IL-1RAPL-2, IL1RAPL2, IL-1RAPL2, IL-1-RAPL-2, IL1RAPL-2IL1RAPL-2-related protein, interleukin 1 receptor 9, interleukin 1 receptor accessory protein-like 2, Three immunoglobulin domain-containing IL-1 receptor-related 1, TIGIRR-1, TIGIRR-1Interleukin-1 receptor 9, X-linked interleukin-1 receptor accessory protein-like 2 | |
| IL1RAPL2 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title