missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL1RAPL2 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB3068-SP
This item is not returnable.
View return policy
Description
IL1RAPL2 Monoclonal specifically detects IL1RAPL2 in Mouse samples. It is validated for Western Blot.
Specifications
| IL1RAPL2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9ERS6 | |
| IL1RAPL2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL1RAPL2, Asn18-Ala358, Accession # Q9ERS6 | |
| 25 μg | |
| Primary | |
| Detects mouse IL1RAPL2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant human (rh) IL1RAPL2 is observed and no cross-reactivity with recombinant mouse (rm) IL-1 R1, rmIL-1 R2, rmIL | |
| Mouse | |
| Purified |
| Western Blot | |
| 320017 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IL-1R10, IL-1R-9, IL1R9IL-1 receptor accessory protein-like 2, IL-1R9IL-1RAPL-2, IL1RAPL2, IL-1RAPL2, IL-1-RAPL-2, IL1RAPL-2IL1RAPL-2-related protein, interleukin 1 receptor 9, interleukin 1 receptor accessory protein-like 2, Three immunoglobulin domain-containing IL-1 receptor-related 1, TIGIRR-1, TIGIRR-1Interleukin-1 receptor 9, X-linked interleukin-1 receptor accessory protein-like 2 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 26280 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction