missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL1RAPL2 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB3068
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
IL1RAPL2 Monoclonal specifically detects IL1RAPL2 in Mouse samples. It is validated for Western Blot.
Spezifikation
| IL1RAPL2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9ERS6 | |
| IL1RAPL2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL1RAPL2, Asn18-Ala358, Accession # Q9ERS6 | |
| 100 μg | |
| Primary | |
| Detects mouse IL1RAPL2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant human (rh) IL1RAPL2 is observed and no cross-reactivity with recombinant mouse (rm) IL-1 R1, rmIL-1 R2, rmIL | |
| Mouse | |
| Purified |
| Western Blot | |
| 320017 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IL-1R10, IL-1R-9, IL1R9IL-1 receptor accessory protein-like 2, IL-1R9IL-1RAPL-2, IL1RAPL2, IL-1RAPL2, IL-1-RAPL-2, IL1RAPL-2IL1RAPL-2-related protein, interleukin 1 receptor 9, interleukin 1 receptor accessory protein-like 2, Three immunoglobulin domain-containing IL-1 receptor-related 1, TIGIRR-1, TIGIRR-1Interleukin-1 receptor 9, X-linked interleukin-1 receptor accessory protein-like 2 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 26280 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur