missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-36 alpha/IL-1F6 Antibody, R&D Systems™

Rat Monoclonal Antibody
£168.00 - £523.00
Specifications
| Antigen | IL-36 alpha/IL-1F6 |
|---|---|
| Clone | 275339 |
| Dilution | Western Blot 1 ug/mL, Blockade of Receptor-ligand Interaction |
| Applications | Western Blot, Blockade of Receptor-ligand Interaction |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18731889
![]() |
R&D Systems
MAB2297-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18189170
|
R&D Systems
MAB2297 |
500 μg |
£523.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-36 alpha/IL-1F6 Monoclonal specifically detects IL-36 alpha/IL-1F6 in Mouse samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.Specifications
| IL-36 alpha/IL-1F6 | |
| Western Blot 1 ug/mL, Blockade of Receptor-ligand Interaction | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FIL1, FIL1 epsilon, FIL1(EPSILON), FIL1E, IL-1 epsilon, IL1(EPSILON), IL1E, IL-1E, IL1F6, IL-1F6, IL-1F6 (FIL-1-epsilon), IL36 alpha, IL36A, interleukin 1 family, member 6 (epsilon), interleukin 1, epsilon, Interleukin 36, Alpha, Interleukin-1 epsilon, interleukin-1 family member 6, Interleukin-36 Alpha | |
| Il1f6 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 275339 | |
| Western Blot, Blockade of Receptor-ligand Interaction | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q9JLA2 | |
| 27179 | |
| E. coli-derived recombinant mouse IL-36 alpha/IL-1F6 Met1-His160 Accession # Q9JLA2 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse IL-36 alpha/IL-1F6 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse (rm) IL-1F5 and no cross-reactivity with recombinant human (rh) rhIL-1 alpha , rhIL-1 beta , rmIL-1F8, rhIL-1F6, 7, 9, 10, or rhIL-18 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title