missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-36 alpha/IL-1F6 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB2297-SP
This item is not returnable.
View return policy
Description
IL-36 alpha/IL-1F6 Monoclonal specifically detects IL-36 alpha/IL-1F6 in Mouse samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.
Specifications
| IL-36 alpha/IL-1F6 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FIL1, FIL1 epsilon, FIL1(EPSILON), FIL1E, IL-1 epsilon, IL1(EPSILON), IL1E, IL-1E, IL1F6, IL-1F6, IL-1F6 (FIL-1-epsilon), IL36 alpha, IL36A, interleukin 1 family, member 6 (epsilon), interleukin 1, epsilon, Interleukin 36, Alpha, Interleukin-1 epsilon, interleukin-1 family member 6, Interleukin-36 Alpha | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 27179 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Blockade of Receptor-ligand Interaction | |
| 275339 | |
| Western Blot 1 ug/mL, Blockade of Receptor-ligand Interaction | |
| Q9JLA2 | |
| Il1f6 | |
| E. coli-derived recombinant mouse IL-36 alpha/IL-1F6 Met1-His160 Accession # Q9JLA2 | |
| 25 μg | |
| Primary | |
| Detects mouse IL-36 alpha/IL-1F6 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse (rm) IL-1F5 and no cross-reactivity with recombinant human (rh) rhIL-1 alpha , rhIL-1 beta , rmIL-1F8, rhIL-1F6, 7, 9, 10, or rhIL-18 is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction