missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-36 alpha/IL-1F6 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB2297
This item is not returnable.
View return policy
Description
IL-36 alpha/IL-1F6 Monoclonal specifically detects IL-36 alpha/IL-1F6 in Mouse samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.
Specifications
| IL-36 alpha/IL-1F6 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FIL1, FIL1 epsilon, FIL1(EPSILON), FIL1E, IL-1 epsilon, IL1(EPSILON), IL1E, IL-1E, IL1F6, IL-1F6, IL-1F6 (FIL-1-epsilon), IL36 alpha, IL36A, interleukin 1 family, member 6 (epsilon), interleukin 1, epsilon, Interleukin 36, Alpha, Interleukin-1 epsilon, interleukin-1 family member 6, Interleukin-36 Alpha | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 27179 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Blockade of Receptor-ligand Interaction | |
| 275339 | |
| Western Blot 1 ug/mL, Blockade of Receptor-ligand Interaction | |
| Q9JLA2 | |
| Il1f6 | |
| E. coli-derived recombinant mouse IL-36 alpha/IL-1F6 Met1-His160 Accession # Q9JLA2 | |
| 500 μg | |
| Primary | |
| Detects mouse IL-36 alpha/IL-1F6 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse (rm) IL-1F5 and no cross-reactivity with recombinant human (rh) rhIL-1 alpha , rhIL-1 beta , rmIL-1F8, rhIL-1F6, 7, 9, 10, or rhIL-18 is observed. | |
| Mouse | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto