missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Dkk-4 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
£157.00 - £432.00
Tekniske data
| Antigen | Dkk-4 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Produktkode | Brand | Quantity | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Quantity | Pris | Antal & tilgængelighed | |||||
18755517
![]() |
R&D Systems
AF3105-SP |
25 μg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18070502
![]() |
R&D Systems
AF3105 |
100 μg |
£432.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivelse
Dkk-4 Polyclonal specifically detects Dkk-4 in Mouse samples. It is validated for Western Blot.Tekniske data
| Dkk-4 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| dickkopf (Xenopus laevis) homolog 4, dickkopf homolog 4 (Xenopus laevis), dickkopf-4, dickkopf-related protein 4, Dkk4, DKK-4, hDkk-4, MGC129562, MGC129563 | |
| DKK4 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q8VEJ3 | |
| 27121 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Dkk-4 Leu19-Ile221 (Arg67Ser, Arg70Ser) Accession # Q8VEJ3 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse Dkk-4 in direct ELISAs and Western blots. In Western blots, less than 5% cross-reactivity with recombinant human Dkk-4, recombinant mouse (rm) Dkk-1, rmDkk-2, and rmDkk-3 is observed. |
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel