Learn More
Mouse Dkk-4 Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody has been used in 2 publications
Brand:  R&D Systems AF3105-SP
 View more versions of this product
Product Code. 18755517
  • £157.00 / 25µg
Description
Dkk-4 Polyclonal specifically detects Dkk-4 in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Dkk-4
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
dickkopf (Xenopus laevis) homolog 4, dickkopf homolog 4 (Xenopus laevis), dickkopf-4, dickkopf-related protein 4, Dkk4, DKK-4, hDkk-4, MGC129562, MGC129563
Goat
Affinity Purified
RUO
27121
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot
LYOPH
Western Blot 0.1 ug/mL
Q8VEJ3
DKK4
Mouse myeloma cell line NS0-derived recombinant mouse Dkk-4 Leu19-Ile221 (Arg67Ser, Arg70Ser) Accession # Q8VEJ3
25 μg
Primary
Detects mouse Dkk-4 in direct ELISAs and Western blots. In Western blots, less than 5% cross-reactivity with recombinant human Dkk-4, recombinant mouse (rm) Dkk-1, rmDkk-2, and rmDkk-3 is observed.