missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Dkk-4 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF3105
This item is not returnable.
View return policy
Description
Dkk-4 Polyclonal specifically detects Dkk-4 in Mouse samples. It is validated for Western Blot.
Specifications
| Dkk-4 | |
| Polyclonal | |
| Unconjugated | |
| Q8VEJ3 | |
| DKK4 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Dkk-4 Leu19-Ile221 (Arg67Ser, Arg70Ser) Accession # Q8VEJ3 | |
| 100 μg | |
| Primary | |
| Detects mouse Dkk-4 in direct ELISAs and Western blots. In Western blots, less than 5% cross-reactivity with recombinant human Dkk-4, recombinant mouse (rm) Dkk-1, rmDkk-2, and rmDkk-3 is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| dickkopf (Xenopus laevis) homolog 4, dickkopf homolog 4 (Xenopus laevis), dickkopf-4, dickkopf-related protein 4, Dkk4, DKK-4, hDkk-4, MGC129562, MGC129563 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 27121 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction