missing translation for 'onlineSavingsMsg'
Learn More

Mouse anti-Human IgG (H+L) Secondary Antibody (4D2D9G8), Novus Biologicals™

Mouse Monoclonal Antibody

£111.00 - £308.00

Specifications

Clone 4D2D9G8
Concentration 1.0 mg/mL
Dilution Western Blot 1:500 - 1:2000, ELISA 1:10000
Applications Western Blot, ELISA
Classification Monoclonal
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18263453
View Documents
Novus Biologicals
NBP1-51523SS
0.025 mL
£111.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18265073
View Documents
Novus Biologicals
NBP1-51523
0.1 mL
£205.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18224705
View Documents
Novus Biologicals
NBP1-51523-0.5ML
0.5 mL
£308.00
0.50mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IgG (H+L) Monoclonal specifically detects IgG (H+L) in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

4D2D9G8
Western Blot 1:500 - 1:2000, ELISA 1:10000
Monoclonal
Purified
RUO
immunoglobulin heavy constant gamma 1 (G1m marker)
IGHG1
IgG1
Protein G purified
Monoclonal anti-Human IgG is derived from a hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse. It is specific for the whole human IgG molecule, and has no cross reactivity with human IgM molecule as determined by an ELISA.
1.0 mg/mL
Western Blot, ELISA
Unconjugated
Mouse
Human
3500
Human IgG was isolated from human sera and purified by chromatography.
Secondary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.