missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgG (H+L) Secondary Antibody (4D2D9G8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-51523
159.80 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
IgG (H+L) Monoclonal antibody specifically detects IgG (H+L) in Human samples. It is validated for Western Blot, ELISA.
Specifications
| IgG (H+L) | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
| IGHG1 | |
| Human IgG was isolated from human sera and purified by chromatography. | |
| 0.1 mL | |
| Secondary | |
| Monoclonal anti-Human IgG is derived from a hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse. It is specific for the whole human IgG molecule, and has no cross reactivity with human IgM molecule as determined by an ELISA. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, ELISA | |
| 4D2D9G8 | |
| Unconjugated | |
| immunoglobulin heavy constant gamma 1 (G1m marker) | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 3500 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction