missing translation for 'onlineSavingsMsg'
Learn More

Mouse anti-Human IgG (H+L) Secondary Antibody (4D2D9G8), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP1-51523-0.5ML

245.65 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.


 View more versions of this product

Product Code. 18224705

  • £308.00 / 0.50mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

IgG (H+L) Monoclonal specifically detects IgG (H+L) in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IgG
Monoclonal
1.0 mg/mL
Western Blot 1:500 - 1:2000, ELISA 1:10000
immunoglobulin heavy constant gamma 1 (G1m marker)
Mouse
Protein G purified
RUO
3500
Human
Purified
Western Blot, ELISA
4D2D9G8
Unconjugated
PBS with 0.03% Sodium Azide
IGHG1
Human IgG was isolated from human sera and purified by chromatography.
0.5 mL
Secondary
Monoclonal anti-Human IgG is derived from a hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse. It is specific for the whole human IgG molecule, and has no cross reactivity with human IgM molecule as determined by an ELISA.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only