missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Monkeypox Virus A35R Recombinant Rabbit Monoclonal Antibody (10I7L8)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£147.00 - £363.00

Specifications

Antigen Monkeypox Virus A35R
Clone 10I7L8
Concentration 1.1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Immunocytochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30283015
View Documents
Invitrogen™
MA558139
20 μL
£147.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30283983
GREENER_CHOICE
View Documents
Invitrogen™
MA558140
100 μL
£363.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Immunogen sequence: RQNQCMSANE AAITDSAVAV AAASSTHRKV ASSTTQYDHK ESCNGLYYQG SCYILHSDYK SFEDAKANCA AESSTLPNKS DVLTTWLIDY VEDTWGSDGN PITKTTSDYQ DSDVSQEVRK YFCT.

The monkeypox virus (MPXV) is the causative agent of the monkeypox disease. As a double-stranded DNA virus member of the genus Orthopoxvirus and the family Poxviridae, MPXV is similar to other pathogens of this group including variola (the causative agent of smallpox), vaccinia, and the cowpox virus. A35R is a Bifunctional EEV Membrane Phosphoglycoprotein (MPXV A35R).
Specifications

Specifications

Monkeypox Virus A35R
1.1 mg/mL
ELISA, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
Recombinant protein (or fragment).
Antibody
10I7L8
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Liquid
IgG
Virus
Monkeypox A35R
Primary
Protein A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.