missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Monkeypox Virus A35R Recombinant Rabbit Monoclonal Antibody (10I7L8)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA558139

 View more versions of this product

Product Code. 30283015

  • £147.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: RQNQCMSANE AAITDSAVAV AAASSTHRKV ASSTTQYDHK ESCNGLYYQG SCYILHSDYK SFEDAKANCA AESSTLPNKS DVLTTWLIDY VEDTWGSDGN PITKTTSDYQ DSDVSQEVRK YFCT.

The monkeypox virus (MPXV) is the causative agent of the monkeypox disease. As a double-stranded DNA virus member of the genus Orthopoxvirus and the family Poxviridae, MPXV is similar to other pathogens of this group including variola (the causative agent of smallpox), vaccinia, and the cowpox virus. A35R is a Bifunctional EEV Membrane Phosphoglycoprotein (MPXV A35R).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Monkeypox Virus A35R
Recombinant Monoclonal
1.1 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
Rabbit
Protein A
RUO
Virus
Antibody
IgG
ELISA, Immunocytochemistry
10I7L8
Unconjugated
Monkeypox A35R
Recombinant protein (or fragment).
20 μL
Primary
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.