missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Monkeypox Virus A35R Recombinant Rabbit Monoclonal Antibody (10I7L8)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558140
This item is not returnable.
View return policy
Description
Immunogen sequence: RQNQCMSANE AAITDSAVAV AAASSTHRKV ASSTTQYDHK ESCNGLYYQG SCYILHSDYK SFEDAKANCA AESSTLPNKS DVLTTWLIDY VEDTWGSDGN PITKTTSDYQ DSDVSQEVRK YFCT.
The monkeypox virus (MPXV) is the causative agent of the monkeypox disease. As a double-stranded DNA virus member of the genus Orthopoxvirus and the family Poxviridae, MPXV is similar to other pathogens of this group including variola (the causative agent of smallpox), vaccinia, and the cowpox virus. A35R is a Bifunctional EEV Membrane Phosphoglycoprotein (MPXV A35R).
Specifications
| Monkeypox Virus A35R | |
| Recombinant Monoclonal | |
| 1.1 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Rabbit | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
| ELISA, Immunocytochemistry | |
| 10I7L8 | |
| Unconjugated | |
| Monkeypox A35R | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction