missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Monkey IL-6R ELISA Kit
Monkey IL-6R quantitates monkey IL-6R in serum, plasma, supernatant.
£530.00 - £3590.00
Specifications
Accession Number | CD126; CD126 antigen; gp80; IL-6 Receptor; il-6 receptor alpha; IL-6 receptor alpha chain; IL-6 receptor subunit alpha; IL6Q; Il6r; IL-6 R; IL-6 R 1; IL-6 R subunit alpha; IL6R1; IL-6 R-1; Il6ra; IL-6 RA; IL-6 R-alpha; IL6RQ; ILN; Interleukin; interleukin 6 receptor; interleukin 6 receptor, alpha; interleukin-6 receptor subunit alpha; Membrane glycoprotein 80; MGC104991 |
---|---|
Assay Range | 12 to 3,000 pg/mL |
Assay Sensitivity | 12 pg/mL |
Conjugate | Biotin |
Product Type | ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17135809
|
Invitrogen™
EP14RBX5 |
5 x 96 Tests |
£2215.00
Each |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
17184933
|
Invitrogen™
EP14RB |
96 Tests |
£530.00
Pack of 96 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
18567824
|
Invitrogen™
EP14RBX10 |
10 x 96 Tests |
£3590.00
Pack of 10 |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Monkey IL-6R quantitates monkey IL-6R in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant monkey IL-6R.
Interleukin 6 receptor (IL6R), also known as CD126 (Cluster of Differentiation 126), is a type I cytokine receptor. This gene encodes a subunit of the interleukin 6 (IL6) receptor complex. Interleukin 6 is a potent pleiotropic cytokine that regulates cell growth and differentiation and plays an important role in the immune response. The IL6 receptor is a protein complex consisting of this protein and interleukin 6 signal transducer (IL6ST/GP130/IL6-beta), a receptor subunit also shared by many other cytokines. Dysregulated production of IL6 and this receptor are implicated in the pathogenesis of many diseases, such as multiple myeloma, autoimmune diseases and prostate cancer. Alternatively spliced transcript variants encoding distinct isoforms have been reported. A pseudogene of this gene is found on chromosome 9.Specifications
CD126; CD126 antigen; gp80; IL-6 Receptor; il-6 receptor alpha; IL-6 receptor alpha chain; IL-6 receptor subunit alpha; IL6Q; Il6r; IL-6 R; IL-6 R 1; IL-6 R subunit alpha; IL6R1; IL-6 R-1; Il6ra; IL-6 RA; IL-6 R-alpha; IL6RQ; ILN; Interleukin; interleukin 6 receptor; interleukin 6 receptor, alpha; interleukin-6 receptor subunit alpha; Membrane glycoprotein 80; MGC104991 | |
12 pg/mL | |
ELISA | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
Research Use Only | |
IL-6 Receptor (CD126) | |
4 hr 45 min |
12 to 3,000 pg/mL | |
Biotin | |
Colorimetric Microplate Reader | |
<10% | |
HRP | |
2°C to 8°C | |
1 hr 20 min |