missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MOG Antibody, R&D Systems™

Goat Polyclonal Antibody
£168.00 - £450.00
Specifications
| Antigen | MOG |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18796487
![]() |
R&D Systems
AF2395-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18081042
![]() |
R&D Systems
AF2395 |
100 μg |
£450.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MOG Polyclonal specifically detects MOG in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| MOG | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| MGC26137, MOG Ig-AluB, MOGIG2, myelin oligodendrocyte glycoprotein, myelin-oligodendrocyte glycoprotein | |
| MOG | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| Q8IYG5 | |
| 4340 | |
| Mouse myeloma cell line NS0-derived recombinant human MOG Gly30-Gly154 (Asp131Ala) Accession # Q8IYG5 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human MOG in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title