missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MOG Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2395-SP
This item is not returnable.
View return policy
Description
MOG Polyclonal specifically detects MOG in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| MOG | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MGC26137, MOG Ig-AluB, MOGIG2, myelin oligodendrocyte glycoprotein, myelin-oligodendrocyte glycoprotein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4340 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Q8IYG5 | |
| MOG | |
| Mouse myeloma cell line NS0-derived recombinant human MOG Gly30-Gly154 (Asp131Ala) Accession # Q8IYG5 | |
| 25 μg | |
| Primary | |
| Detects human MOG in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction