missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MOG Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2395
This item is not returnable.
View return policy
Description
MOG Polyclonal specifically detects MOG in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| MOG | |
| Polyclonal | |
| Unconjugated | |
| Q8IYG5 | |
| MOG | |
| Mouse myeloma cell line NS0-derived recombinant human MOG Gly30-Gly154 (Asp131Ala) Accession # Q8IYG5 | |
| 100 μg | |
| Primary | |
| Detects human MOG in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| MGC26137, MOG Ig-AluB, MOGIG2, myelin oligodendrocyte glycoprotein, myelin-oligodendrocyte glycoprotein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4340 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction