missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP2 / Collagenase Type IV A Mouse, Clone: MMP2/1501, enQuireBio™
Mouse Monoclonal Antibody
£181.83 - £395.00
Specifications
Antigen | MMP2 / Collagenase Type IV A |
---|---|
Clone | MMP2/1501 |
Concentration | 200μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15940488
|
EnquireBio
4313-MSM1-P1 |
100 μg |
£395.00
100µg |
Estimated Shipment: 04-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15930488
|
EnquireBio
4313-MSM1-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
MMP2 / Collagenase Type IV A | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
4313 | |
Recombinant human MMP2 protein fragment (aa444-575) | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
72kDa (Pro) ;63kDa (cleaved) |
MMP2/1501 | |
Western Blot | |
Unconjugated | |
RUO | |
NP_001289438 | |
MMP2 | |
IgG1 | |
Protein A/G Purified | |
Cytoplasmic and Cell Surface | |
U-138 MG or U-87 MG cells. Placenta |