missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP2 / Collagenase Type IV A Mouse, Clone: MMP2/1501, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 4313-MSM1-P1
Additional Details : Weight : 0.01000kg
Specifications
MMP2 / Collagenase Type IV A | |
Cytoplasmic and Cell Surface | |
MMP2/1501 | |
Unconjugated | |
NP_001289438 | |
Mouse | |
72kDa (Pro) ;63kDa (cleaved) | |
Protein A/G Purified | |
RUO | |
4313 | |
IgG1 |
Western Blot | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
MMP2 | |
Recombinant human MMP2 protein fragment (aa444-575) | |
U-138 MG or U-87 MG cells. Placenta | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |