missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-7 Rabbit anti-Human, Mouse, Rat, Clone: 9D8S10, Novus Biologicals™
Rabbit Monoclonal Antibody
£167.00 - £416.00
Specifications
| Antigen | MMP-7 |
|---|---|
| Clone | 9D8S10 |
| Dilution | Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18364972
|
Bio-Techne
NBP3-16067-20UL |
20 μg |
£167.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18046734
|
Novus Biologicals
NBP3-16067-100UL |
100 μg |
£416.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MMP-7 Monoclonal antibody specifically detects MMP-7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| MMP-7 | |
| Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 3.4.24, EC 3.4.24.23, matrilysin, matrin, matrix metallopeptidase 7 (matrilysin, uterine), matrix metalloproteinase 7 (matrilysin, uterine), Matrix metalloproteinase-7, MMP-7, MPSL1, PUMP1, PUMP-1, Pump-1 protease, uterine matrilysin, Uterine metalloproteinase | |
| Recombinant protein of human MMP-7. | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 9D8S10 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Wnt Signaling Pathway | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 4316 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title