missing translation for 'onlineSavingsMsg'
Learn More

MMP-7 Rabbit anti-Human, Mouse, Rat, Clone: 9D8S10, Novus Biologicals™

Product Code. 18364972
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18364972 20 μg 20µL
18046734 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18364972 Supplier Bio-Techne Supplier No. NBP31606720UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

MMP-7 Monoclonal antibody specifically detects MMP-7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP-7
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 9D8S10
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias EC 3.4.24, EC 3.4.24.23, matrilysin, matrin, matrix metallopeptidase 7 (matrilysin, uterine), matrix metalloproteinase 7 (matrilysin, uterine), Matrix metalloproteinase-7, MMP-7, MPSL1, PUMP1, PUMP-1, Pump-1 protease, uterine matrilysin, Uterine metalloproteinase
Host Species Rabbit
Immunogen Recombinant protein of human MMP-7.
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 4316
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.