missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-14/MT1-MMP Antibody (114-6G6), Novus Biologicals™
Mouse Monoclonal Antibody
£171.00 - £538.00
Specifications
| Antigen | MMP-14/MT1-MMP |
|---|---|
| Clone | 114-6G6 |
| Concentration | 2 mg/mL |
| Dilution | Western Blot 8-20 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry-Paraffin 1:10-1:500 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18162305
|
Novus Biologicals
NBP2-29699-0.025MG |
0.025 mg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18173753
|
Novus Biologicals
NBP2-29699 |
0.1 mg |
£538.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MMP-14/MT1-MMP Monoclonal specifically detects MMP-14/MT1-MMP in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| MMP-14/MT1-MMP | |
| 2 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| Human | |
| EC 3.4.24, EC 3.4.24.80, matrix metallopeptidase 14 (membrane-inserted), matrix metalloproteinase 14 (membrane-inserted), matrix metalloproteinase-14, membrane type 1 metalloprotease, Membrane-type matrix metalloproteinase 1, Membrane-type-1 matrix metalloproteinase, MMP-14, MMP-X1, MT1MMP, MT1-MMPMTMMP1, MT-MMP 1 | |
| MMP14 | |
| IgG1 κ | |
| Affinity Purified | |
| The antibody specifically reacts with human MT1-MMP. This is a purified mouse monoclonal antibody to an oligopeptide of REVPYAYIREGHEK (residue 160-173) on human membrane-type 1 matrix metalloproteinase (human MT1-MMP) and does not react with mouse or rat. However, this sequence is identical in mouse and rat MT1-MMP. For unknown reasons this antibody does not react on westerns to CC1043. |
| 114-6G6 | |
| Western Blot 8-20 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Monoclonal | |
| Mouse | |
| Cancer, Extracellular Matrix | |
| P50281 | |
| 4323 | |
| This antibody is specific to MMP-14 | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title