missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-14/MT1-MMP Antibody (114-6G6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-29699-0.025MG
This item is not returnable.
View return policy
Description
MMP-14/MT1-MMP Monoclonal specifically detects MMP-14/MT1-MMP in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| MMP-14/MT1-MMP | |
| Monoclonal | |
| 2 mg/mL | |
| Western Blot 8-20 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry-Paraffin 1:10-1:500 | |
| P50281 | |
| MMP14 | |
| This antibody is specific to MMP-14 | |
| 0.025 mg | |
| Cancer, Extracellular Matrix | |
| 4323 | |
| Human | |
| IgG1 κ |
| Immunohistochemistry (Paraffin), Western Blot, ELISA | |
| 114-6G6 | |
| Unconjugated | |
| 0.1M Sodium Phosphate (pH 7.0) and 2% BSA (protease free) with No Preservative | |
| EC 3.4.24, EC 3.4.24.80, matrix metallopeptidase 14 (membrane-inserted), matrix metalloproteinase 14 (membrane-inserted), matrix metalloproteinase-14, membrane type 1 metalloprotease, Membrane-type matrix metalloproteinase 1, Membrane-type-1 matrix metalloproteinase, MMP-14, MMP-X1, MT1MMP, MT1-MMPMTMMP1, MT-MMP 1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The antibody specifically reacts with human MT1-MMP. This is a purified mouse monoclonal antibody to an oligopeptide of REVPYAYIREGHEK (residue 160-173) on human membrane-type 1 matrix metalloproteinase (human MT1-MMP) and does not react with mouse or rat. However, this sequence is identical in mouse and rat MT1-MMP. For unknown reasons this antibody does not react on westerns to CC1043. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction