Learn More
Methyl 4-formylbenzoate, 98+%, Thermo Scientific Chemicals
£34.00 - £92.10
Chemical Identifiers
CAS | 1571-08-0 |
---|---|
Molecular Formula | C9H8O3 |
Molecular Weight (g/mol) | 164.16 |
MDL Number | MFCD00006950 |
InChI Key | FEIOASZZURHTHB-UHFFFAOYSA-N |
Synonym | methyl p-formylbenzoate, 4-carbomethoxybenzaldehyde, methyl terephthalaldehydate, 4-methoxycarbonyl benzaldehyde, benzoic acid, 4-formyl-, methyl ester, p-carbomethoxybenzaldehyde, methyl terephthaldehydate, methyl4-formylbenzoate, 4-formylbenzoic acid methyl ester, methyl-p-formyl benzoate |
PubChem CID | 15294 |
IUPAC Name | methyl 4-formylbenzoate |
SMILES | COC(=O)C1=CC=C(C=O)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11421097
|
Thermo Scientific Alfa Aesar
B24091.06 |
5 g |
£34.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11431097
|
Thermo Scientific Alfa Aesar
B24091.14 |
25 g |
£61.80
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11441097
|
Thermo Scientific Alfa Aesar
B24091.22 |
100 g |
£92.10
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Methyl 4-formylbenzoate is used in the preparation of dimethyl terephthalate. It is also used as an active pharmaceutical ingredient intermediate and fluorescent brightener intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl 4-formylbenzoate is used in the preparation of dimethyl terephthalate. It is also used as an active pharmaceutical ingredient intermediate and fluorescent brightener intermediate.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
1571-08-0 | |
164.16 | |
FEIOASZZURHTHB-UHFFFAOYSA-N | |
15294 | |
COC(=O)C1=CC=C(C=O)C=C1 |
C9H8O3 | |
MFCD00006950 | |
methyl p-formylbenzoate, 4-carbomethoxybenzaldehyde, methyl terephthalaldehydate, 4-methoxycarbonyl benzaldehyde, benzoic acid, 4-formyl-, methyl ester, p-carbomethoxybenzaldehyde, methyl terephthaldehydate, methyl4-formylbenzoate, 4-formylbenzoic acid methyl ester, methyl-p-formyl benzoate | |
methyl 4-formylbenzoate |
Specifications
1571-08-0 | |
1.2 | |
128°C (262°F) | |
MFCD00006950 | |
Air Sensitive | |
Insoluble in water. | |
COC(=O)C1=CC=C(C=O)C=C1 | |
164.16 | |
164.16 | |
Methyl 4-formylbenzoate |
59°C to 63°C | |
264°C to 266°C | |
C9H8O3 | |
775895 | |
methyl p-formylbenzoate, 4-carbomethoxybenzaldehyde, methyl terephthalaldehydate, 4-methoxycarbonyl benzaldehyde, benzoic acid, 4-formyl-, methyl ester, p-carbomethoxybenzaldehyde, methyl terephthaldehydate, methyl4-formylbenzoate, 4-formylbenzoic acid methyl ester, methyl-p-formyl benzoate | |
FEIOASZZURHTHB-UHFFFAOYSA-N | |
methyl 4-formylbenzoate | |
15294 | |
≥98% |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 216-385-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only