missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-formylbenzoate, 98+%

Product Code. 11421097
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11421097 5 g 5g
11431097 25 g 25g
11441097 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11421097 Supplier Thermo Scientific Alfa Aesar Supplier No. B24091.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1571-08-0 | C9H8O3 | 164.16 g/mol

Methyl 4-formylbenzoate is used in the preparation of dimethyl terephthalate. It is also used as an active pharmaceutical ingredient intermediate and fluorescent brightener intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-formylbenzoate is used in the preparation of dimethyl terephthalate. It is also used as an active pharmaceutical ingredient intermediate and fluorescent brightener intermediate.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1571-08-0
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00006950
InChI Key FEIOASZZURHTHB-UHFFFAOYSA-N
Synonym methyl p-formylbenzoate, 4-carbomethoxybenzaldehyde, methyl terephthalaldehydate, 4-methoxycarbonyl benzaldehyde, benzoic acid, 4-formyl-, methyl ester, p-carbomethoxybenzaldehyde, methyl terephthaldehydate, methyl4-formylbenzoate, 4-formylbenzoic acid methyl ester, methyl-p-formyl benzoate
PubChem CID 15294
IUPAC Name methyl 4-formylbenzoate
SMILES COC(=O)C1=CC=C(C=O)C=C1

Specifikationer

Melting Point 59°C to 63°C
Density 1.2
Boiling Point 264°C to 266°C
Flash Point 128°C (262°F)
Quantity 5 g
Beilstein 775895
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 164.16
Percent Purity ≥98%
Chemical Name or Material Methyl 4-formylbenzoate
Visa mer Visa mindre

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.